missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine 95.0+%, TCI America™

Catalog No. B5217200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5217200MG 200 mg
B52171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5217200MG Supplier TCI America Supplier No. B5217200MG
Only null left

Chemical Identifiers

CAS 1153949-11-1
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.234
MDL Number MFCD12761241
InChI Key BESFCRTTXQYNBW-UHFFFAOYSA-N
Synonym 1-Boc-3-(cyanomethylene)azetidine, tert-Butyl 3-(Cyanomethylene)azetidine-1-carboxylate, 3-(Cyanomethylene)azetidine-1-carboxylic Acid tert-Butyl Ester
PubChem CID 45789216
IUPAC Name tert-butyl 3-(cyanomethylidene)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(=CC#N)C1

Specifications

Color White-Yellow
UN Number 3439
Quantity 200 mg
Formula Weight 194.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine
TSCA No
Recommended Storage Refrigerator