missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-3-azetidinemethanol 98.0+%, TCI America™

Catalog No. B39921G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B39921G Supplier TCI America Supplier No. B39921G

Chemical Identifiers

CAS 142253-56-3
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD06656141
InChI Key HXRDRJKAEYHOBB-UHFFFAOYSA-N
Synonym tert-butyl 3-hydroxymethyl azetidine-1-carboxylate, 1-boc-azetidine-3-yl-methanol, 1-boc-azetidine-3-yl methanol, 1-boc-azetidine-3-ylmethanol, 1-boc-3-hydroxymethyl azetidine, 1-boc-3-azetidinemethanol, 1-tert-butoxycarbonyl-3-azetidinemethanol, 1-azetidinecarboxylic acid, 3-hydroxymethyl-, 1,1-dimethylethyl ester, tert-butyl3-hydroxymethyl azetidine-1-carboxylate, 3-hydroxymethyl-azetidine-1-carboxylic acid tert-butyl ester
PubChem CID 10583745
IUPAC Name tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CO)C1

Specifications

Melting Point 55°C
Boiling Point 273°C
Color White-Yellow
Quantity 1 g
Formula Weight 187.24
Percent Purity ≥98.0% (GC)
Physical Form Crystal/Lump
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-azetidinemethanol