missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)-2-pyrroleboronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Melting Point | 102°C |
| Color | White |
| Quantity | 1 g |
| Synonym | n-boc-2-pyrroleboronic acid, n-boc-pyrrole-2-boronic acid, 1-boc-pyrrole-2-boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-ylboronic acid, 1-tert-butoxycarbonyl pyrrole-2-boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid, 1-t-butoxycarbonyl pyrrole-2-boronic acid, boc-pyrrole-2-boronic acid, 1-boc-2-pyrrolylboronic acid, 1-tert-butoxy carbonyl-1h-pyrrol-2-yl boronic acid |
| SMILES | B(C1=CC=CN1C(=O)OC(C)(C)C)(O)O |
| IUPAC Name | [1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrol-2-yl]boronic acid |
| Molecular Weight (g/mol) | 211.024 |
| Formula Weight | 211.02 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-(tert-Butoxycarbonyl)-2-pyrroleboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |