missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butoxycarbonyl-1,2,4-triazole 97.0+%, TCI America™

Catalog No. b196925g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B196925G 25 g
B19695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B196925G Supplier TCI America Supplier No. B196925G
Only null left

Chemical Identifiers

CAS 41864-24-8
Molecular Formula C7H11N3O2
Molecular Weight (g/mol) 169.18
MDL Number MFCD00042665
InChI Key XWUFRDJEUOAMNW-UHFFFAOYSA-N
Synonym 1-Boc-1,2,4-triazole
PubChem CID 3716936
IUPAC Name tert-butyl 1H-1,2,4-triazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=NC=N1

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 75°C
Quantity 25 g
Formula Weight 169.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butoxycarbonyl-1,2,4-triazole
TSCA No
Recommended Storage Refrigerator