missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-sec-Butyl-4-nitrobenzene 97.0+%, TCI America™
$234.87 - $631.46
Chemical Identifiers
| CAS | 4237-40-5 |
|---|---|
| Molecular Formula | C10H13NO2 |
| Molecular Weight (g/mol) | 179.22 |
| MDL Number | MFCD00130025 |
| InChI Key | IQRMCUYGEZOTSV-UHFFFAOYNA-N |
| PubChem CID | 107249 |
| IUPAC Name | 1-(butan-2-yl)-4-nitrobenzene |
| SMILES | CCC(C)C1=CC=C(C=C1)[N+]([O-])=O |
Chemical Identifiers
| 4237-40-5 | |
| 179.22 | |
| IQRMCUYGEZOTSV-UHFFFAOYNA-N | |
| 1-(butan-2-yl)-4-nitrobenzene |
| C10H13NO2 | |
| MFCD00130025 | |
| 107249 | |
| CCC(C)C1=CC=C(C=C1)[N+]([O-])=O |
Specifications
| 4237-40-5 | |
| C10H13NO2 | |
| 5 g | |
| CCC(C)C1=CC=C(C=C1)[N+]([O-])=O | |
| 179.22 | |
| 179.22 | |
| Liquid |
| 144°C | |
| MFCD00130025 | |
| IQRMCUYGEZOTSV-UHFFFAOYNA-N | |
| 1-(butan-2-yl)-4-nitrobenzene | |
| 107249 | |
| ≥97.0% (GC) | |
| 1-sec-Butyl-4-nitrobenzene |
Safety and Handling
TSCA : Yes