missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-sec-Butyl-4-nitrobenzene 97.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 144°C |
| Quantity | 5 g |
| Formula Weight | 179.22 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-sec-Butyl-4-nitrobenzene |
Safety and Handling
| TSCA | Yes |