missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pyrrolidino-1-cyclopentene 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 93°C |
| Quantity | 5 g |
| UN Number | 1993 |
| Formula Weight | 137.23 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Pyrrolidino-1-cyclopentene |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Refrigerator |