missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrrolidinecarbonyl chloride, 97%

Catalog No. AAL1339603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1339603 1 g
AAL1339606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1339603 Supplier Thermo Scientific Chemicals Supplier No. L1339603
Only null left

CAS: 1192-63-8 | C5H8ClNO | 133.575 g/mol

1-Pyrrolidinecarbonyl chloride, is used in the synthesis of (S)-4-(2-(((benzyloxy)carbonyl)amino)-3-methoxy-3-oxopropyl)phenyl pyrrolidine-1-carboxylate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrrolidinecarbonyl chloride, is used in the synthesis of (S)-4-(2-(((benzyloxy)carbonyl)amino)-3-methoxy-3-oxopropyl)phenyl pyrrolidine-1-carboxylate

Solubility
It reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.

Chemical Identifiers

CAS 1192-63-8
Molecular Formula C5H8ClNO
Molecular Weight (g/mol) 133.575
MDL Number MFCD00051321
InChI Key XACWJIQLDLUFSR-UHFFFAOYSA-N
Synonym 1-pyrrolidinecarbonyl chloride, pyrrolidinecarbonyl chloride, 1-pyrrolidinecarbonylchloride, pyrrolidine-1-carbonylchloride, pyrrolidinocarbonyl chloride, 1pyrrolidinecarbonyl chloride, pyrrolidine carbonyl chloride, acmc-1bo54, 1-pyrrolidine carbonylchloride, 1-pyrolidine carbonyl chloride
PubChem CID 70937
IUPAC Name pyrrolidine-1-carbonyl chloride
SMILES C1CCN(C1)C(=O)Cl

Specifications

Boiling Point 75°C (1mmHg)
Refractive Index 1.494
UN Number UN3265
Beilstein 110281
Solubility Information It reacts with water.
Density 1.209
Quantity 1 g
Flash Point 97°C (206°F)
Formula Weight 133.58
Sensitivity Moisture sensitive
Percent Purity 97%
Chemical Name or Material 1-Pyrrolidinecarbonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 214-758-7
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only