missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrrolidinecarbodithioic acid ammonium salt, 98%

Catalog No. AAB2373114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2373114 25 g
AAB2373122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2373114 Supplier Thermo Scientific Chemicals Supplier No. B2373114

CAS: 5108-96-3 | C5H8NS2- | 146.246 g/mol

1-Pyrrolidinecarbodithioic acid ammonium salt is useful for biochemical applications including metal chelation. It is used to prevent the induction of nitric oxide synthase. It is also essential to inhibit apoptosis in leukemia HL-60 cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrrolidinecarbodithioic acid ammonium salt is useful for biochemical applications including metal chelation. It is used to prevent the induction of nitric oxide synthase. It is also essential to inhibit apoptosis in leukemia HL-60 cells.

Solubility
Soluble in water.

Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 5108-96-3
Molecular Formula C5H8NS2-
Molecular Weight (g/mol) 146.246
MDL Number MFCD00012720
InChI Key VSWDORGPIHIGNW-UHFFFAOYSA-M
Synonym ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium
PubChem CID 4311638
IUPAC Name pyrrolidine-1-carbodithioate
SMILES C1CCN(C1)C(=S)[S-]

Specifications

Melting Point 150°C to 155°C
Quantity 25 g
Beilstein 3730472
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 164.29
Percent Purity 98%
Chemical Name or Material 1-Pyrrolidinecarbodithioic acid ammonium salt
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-834-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only