missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrimidin-2-yl-piperidine-4-carboxylic acid, Thermo Scientific™

Chemical Identifiers

CAS 303144-44-7
Molecular Formula C10H12N3O2
Molecular Weight (g/mol) 206.23
MDL Number MFCD00664474
InChI Key DYVXURZASBPYFR-UHFFFAOYSA-M
Synonym 1-pyrimidin-2-yl piperidine-4-carboxylic acid, 1-pyrimidin-2-yl-piperidine-4-carboxylic acid, 1-2-pyrimidinyl piperidine-4-carboxylic acid, 1-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-2-pyrimidyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-2-pyrimidinyl, acmc-1agno, 4-piperidinecarboxylicacid, 1-2-pyrimidinyl
PubChem CID 2767267
IUPAC Name 1-pyrimidin-2-ylpiperidine-4-carboxylic acid
SMILES [O-]C(=O)C1CCN(CC1)C1=NC=CC=N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC20701DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC20701DE
View Documents
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC20701EA
View Documents
N/A 10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

303144-44-7
206.23
DYVXURZASBPYFR-UHFFFAOYSA-M
2767267
[O-]C(=O)C1CCN(CC1)C1=NC=CC=N1
C10H12N3O2
MFCD00664474
1-pyrimidin-2-yl piperidine-4-carboxylic acid, 1-pyrimidin-2-yl-piperidine-4-carboxylic acid, 1-2-pyrimidinyl piperidine-4-carboxylic acid, 1-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-2-pyrimidyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-2-pyrimidinyl, acmc-1agno, 4-piperidinecarboxylicacid, 1-2-pyrimidinyl
1-pyrimidin-2-ylpiperidine-4-carboxylic acid

Specifications

303144-44-7
C10H12N3O2
1-pyrimidin-2-yl piperidine-4-carboxylic acid, 1-pyrimidin-2-yl-piperidine-4-carboxylic acid, 1-2-pyrimidinyl piperidine-4-carboxylic acid, 1-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-2-pyrimidyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-2-pyrimidinyl, acmc-1agno, 4-piperidinecarboxylicacid, 1-2-pyrimidinyl
[O-]C(=O)C1CCN(CC1)C1=NC=CC=N1
206.23
207.23
1-Pyrimidin-2-yl-piperidine-4-carboxylic acid
Amber glass bottle
MFCD00664474
DYVXURZASBPYFR-UHFFFAOYSA-M
1-pyrimidin-2-ylpiperidine-4-carboxylic acid
2767267
97%
Videos
SDS
Documents

Documents