missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pyrenecarboxylic Acid 97.0+%, TCI America™
$136.08 - $408.22
Chemical Identifiers
| CAS | 19694-02-1 |
|---|---|
| Molecular Formula | C17H10O2 |
| Molecular Weight (g/mol) | 246.27 |
| MDL Number | MFCD00192424 |
| InChI Key | HYISVWRHTUCNCS-UHFFFAOYSA-N |
| PubChem CID | 125395 |
| IUPAC Name | pyrene-1-carboxylic acid |
| SMILES | OC(=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 |
Chemical Identifiers
| 19694-02-1 | |
| 246.27 | |
| HYISVWRHTUCNCS-UHFFFAOYSA-N | |
| pyrene-1-carboxylic acid |
| C17H10O2 | |
| MFCD00192424 | |
| 125395 | |
| OC(=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 |
Specifications
| 19694-02-1 | |
| C17H10O2 | |
| 1 g | |
| OC(=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 | |
| 246.27 | |
| 246.27 | |
| Crystalline Powder |
| 274°C | |
| MFCD00192424 | |
| HYISVWRHTUCNCS-UHFFFAOYSA-N | |
| pyrene-1-carboxylic acid | |
| 125395 | |
| ≥97.0% (GC,T) | |
| 1-Pyrenecarboxylic Acid |
Safety and Handling
TSCA : No