missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pyrenecarboxylic Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 274°C |
| Quantity | 5 g |
| Formula Weight | 246.27 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Pyrenecarboxylic Acid |
Safety and Handling
| TSCA | No |