missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenecarboxylic Acid 97.0+%, TCI America™

Catalog No. P16871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16871G 1 g
P16875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16871G Supplier TCI America Supplier No. P16871G
Only null left

Chemical Identifiers

CAS 19694-02-1
Molecular Formula C17H10O2
Molecular Weight (g/mol) 246.27
MDL Number MFCD00192424
InChI Key HYISVWRHTUCNCS-UHFFFAOYSA-N
PubChem CID 125395
IUPAC Name pyrene-1-carboxylic acid
SMILES OC(=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 274°C
Quantity 1 g
Formula Weight 246.27
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Pyrenecarboxylic Acid