missing translation for 'onlineSavingsMsg'
Learn More

1-Propynyltri-n-butyltin, 96%

Catalog No. AAL2020103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2020103 1 g
AAL2020106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL2020103 Supplier Thermo Scientific Chemicals Supplier No. L2020103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 64099-82-7 | C15H30Sn | 329.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64099-82-7
Molecular Formula C15H30Sn
Molecular Weight (g/mol) 329.115
MDL Number MFCD01863650
InChI Key KCQJLTOSSVXOCC-UHFFFAOYSA-N
Synonym tributyl 1-propynyl tin, tributyl prop-1-yn-1-yl stannane, tributyl prop-1-ynyl stannane, tributylpropynylstannane, 1-propynyltri-n-butyltin, stannane, tributyl-1-propynyl, tributyl-1-propyn-1-ylstannane, tributyl-prop-1-ynyl-stannane, propynyltributyltin;, ch3ccsnbu3
PubChem CID 2733165
IUPAC Name tributyl(prop-1-ynyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C#CC

Specifications

Density 1.082
Boiling Point 277°C
Flash Point >110°C (230°F)
Refractive Index 1.483
Linear Formula H3CC≡CSn[(CH2)3CH3]3
Quantity 1 g
UN Number UN2788
Formula Weight 329.12
Percent Purity 96%
Chemical Name or Material 1-Propynyltri-n-butyltin
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H300-H311-H372-H315-H319
Fatal if swallowed.
Toxic in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only