missing translation for 'onlineSavingsMsg'
Learn More

1-Propylpyridinium Chloride 98.0+%, TCI America™

Catalog No. P13935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13935G Supplier TCI America Supplier No. P13935G

Crystalline powder with ≥98.0% purity (titration).

Chemical Identifiers

CAS 23271-47-8
Molecular Formula C8H12ClN
Molecular Weight (g/mol) 157.641
MDL Number MFCD06797153
InChI Key IBOIUWAYPMADRC-UHFFFAOYSA-M
PubChem CID 13561212
IUPAC Name 1-propylpyridin-1-ium;chloride
SMILES CCC[N+]1=CC=CC=C1.[Cl-]

Specifications

Quantity 5 g
Formula Weight 157.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Propylpyridinium Chloride