missing translation for 'onlineSavingsMsg'
Learn More

1-Propylimidazole 98.0+%, TCI America™

Catalog No. P199125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P199125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P199125G Supplier TCI America Supplier No. P199125G
Only null left

Chemical Identifiers

CAS 35203-44-2
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00060494
InChI Key IYVYLVCVXXCYRI-UHFFFAOYSA-N
PubChem CID 118785
IUPAC Name 1-propylimidazole
SMILES CCCN1C=CN=C1

Specifications

Color Yellow
Boiling Point 94°C
Quantity 25 g
Formula Weight 110.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Propylimidazole