missing translation for 'onlineSavingsMsg'
Learn More

1-Propylboronic acid, 98%

Catalog No. AAL1996514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1996514 25 g
AAL1996503 1 g
AAL1996506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1996514 Supplier Thermo Scientific Chemicals Supplier No. L1996514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17745-45-8 | C3H9BO2 | 87.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17745-45-8
Molecular Formula C3H9BO2
Molecular Weight (g/mol) 87.91
MDL Number MFCD01074564
InChI Key JAQOMSTTXPGKTN-UHFFFAOYSA-N
Synonym n-propylboronic acid, 1-propaneboronic acid, propaneboronic acid, boronic acid, propyl, 1-propylboronic acid, 1-propaneboronic, n-propylboronc acd, propyl boronic acid, pubchem6019
PubChem CID 351065
IUPAC Name propylboronic acid
SMILES CCCB(O)O

Specifications

Melting Point 105°C to 107°C
Quantity 25 g
Beilstein 1731884
Sensitivity Air sensitive; Hygroscopic
Formula Weight 87.91
Percent Purity 98%
Chemical Name or Material 1-Propylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only