missing translation for 'onlineSavingsMsg'
Learn More

1-Propoxy-2-propanol (contains 2-Isopropoxy-1-propanol) 93.0+%, TCI America™

Catalog No. p-7135373
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P204225ML 25 mL
P2042500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P204225ML Supplier TCI America Supplier No. P204225ML

Chemical Identifiers

CAS 1569-01-3
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00192420
InChI Key FENFUOGYJVOCRY-UHFFFAOYNA-N
Synonym Propylene Glycol Monopropyl Ether
PubChem CID 15286
IUPAC Name 1-propoxypropan-2-ol
SMILES CCCOCC(C)O

Specifications

Color Yellow
Boiling Point 149°C
Quantity 25 mL
UN Number 1993
Formula Weight 118.18
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Propoxy-2-propanol (contains 2-Isopropoxy-1-propanol)
EINECSNumber (2)-2424&(7)-0097
RTECSNumber UB9351000
TSCA Yes