missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanol 99.5+%, TCI America™
$44.73 - $52.19
Chemical Identifiers
| CAS | 71-23-8 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.10 |
| MDL Number | MFCD00002941 |
| InChI Key | BDERNNFJNOPAEC-UHFFFAOYSA-N |
| Synonym | 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol |
| PubChem CID | 1031 |
| ChEBI | CHEBI:28831 |
| IUPAC Name | propan-1-ol |
| SMILES | CCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
P049125ML
|
TCI America
P049125ML |
25 mL |
Each for $44.73
|
|
|||||
|
P0491500ML
|
TCI America
P0491500ML |
500 mL |
Each for $52.19
|
|
|||||
Chemical Identifiers
| 71-23-8 | |
| 60.10 | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| 1031 | |
| propan-1-ol |
| C3H8O | |
| MFCD00002941 | |
| 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
| CHEBI:28831 | |
| CCCO |
Specifications
| 71-23-8 | |
| Colorless | |
| C3H8O | |
| 25 mL | |
| 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
| CCCO | |
| 60.10 | |
| CHEBI:28831 | |
| ≥99.5% (GC) | |
| 1-Propanol |
| -127°C | |
| 97°C | |
| MFCD00002941 | |
| 1274 | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| propan-1-ol | |
| 1031 | |
| 60.10 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0207
RTECSNumber : UH8225000
TSCA : Yes