missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, specified according to the requirements of Ph.Eur., Thermo Scientific™

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC446270010
SDS View Documents
Thermo Scientific Chemicals
446270010
1L Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC446270025
SDS View Documents
Thermo Scientific Chemicals
446270025
2.5L Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO

Specifications

71-23-8
0.01 max. at 310nm, 0.02 max. at 290nm, 0.03 max. at 270nm, 0.1 max. at 250nm, 0.3 max. at 230nm
97.0°C
Authentic
C3H8O
CH3CH2CH2OH
1L
15,7955
Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
CCCO
60.10
2.2 mPa.s (20°C)
60.1
0.3 (Total impurities) max
passes test
0.8000g/mL
15°C
Glass bottle
1.3840 to 1.3860
MFCD00002941
0.8
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
CHEBI:28831
≥99.7% (GC)
1-Propanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 200-746-9

SDS
Documents

Documents

RUO – Research Use Only