missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanol, purum, ≥99% (GC), Honeywell™
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Melting Point | -127° |
| Density | 0.804g/mL at 25°C |
| Quantity | 20 kg |
| Explosion Limit | 0.137 |
| Flash Point | 22°C (71.6°F) |
| Ignition Point | 700°F |
| Packaging | Metal Drum |
| Physical Form | Liquid |
| Linear Formula | CH3CH2CH2OH |
| Vapor Pressure | 10mmHg (147°C), 14.9mmHg (20°C) |
| Show More |
Safety and Handling
| Hazard Statement | P210-P261-P280-P305 + P351 + P338 |
| Precautionary Statement | H225-H318-H336 |
| DOTInformation | Class 3 - PG 2 - n-Propanol |
| EINECSNumber | 200-746-9 |
| RTECSNumber | UH8225000 |