missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, purum, ≥99% (GC), Solstice

Catalog No. 60045522
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 kg
4 x 2.5 L
6 x 1 L
160 kg
Packaging:
Metal Drum
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045522 1 L Poly Bottle
60045523 2.5 L Poly Bottle
60045526 6 x 1 L Poly Bottle
60045525 4 x 2.5 L Poly Bottle
60045524 20 kg Metal Drum
60045521 160 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60045522 Supplier Solstice Supplier No. 241351L

≥99% (GC), Purum

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127°
Density 0.804g/mL at 25°C
Quantity 1 L
Explosion Limit 0.137
Flash Point 22°C (71.6°F)
Ignition Point 700°F
Packaging Poly Bottle
Physical Form Liquid
Linear Formula CH3CH2CH2OH
Vapor Pressure 10mmHg (147°C), 14.9mmHg (20°C)
UN Number UN1274
Beilstein 1098242
Boiling Point 97°
Formula Weight 60.1g/mol
Percent Purity ≥99% (GC)
Grade purum
Chemical Name or Material 1-Propanol
Show More Show Less
Hazard Statement P210-P261-P280-P305 + P351 + P338
Precautionary Statement H225-H318-H336
DOTInformation Class 3 - PG 2 - n-Propanol
EINECSNumber 200-746-9
RTECSNumber UH8225000