missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanol, purum, ≥99% (GC), Honeywell™
≥99% (GC), Purum
Supplier: Honeywell Chemicals 24135160KG
Description
- ≥95% for general laboratory use.
Specifications
| 1-Propanol | |
| 71-23-8 | |
| 97° | |
| 0.137 | |
| 700°F | |
| Liquid | |
| CH3CH2CH2OH | |
| 10mmHg (147°C), 14.9mmHg (20°C) | |
| 1098242 | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| propan-1-ol | |
| 1031 | |
| 60.1g/mol | |
| purum |
| -127° | |
| 0.804g/mL at 25°C | |
| 160 kg | |
| 22°C (71.6°F) | |
| Metal Drum | |
| C3H8O | |
| MFCD00002941 | |
| UN1274 | |
| 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
| CCCO | |
| 60.10 | |
| CHEBI:28831 | |
| ≥99% (GC) |
Chemical Identifiers
| 71-23-8 | |
| 60.10 | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| 1031 | |
| propan-1-ol |
| C3H8O | |
| MFCD00002941 | |
| 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
| CHEBI:28831 | |
| CCCO |
Safety and Handling
P210-P261-P280-P305 + P351 + P338
H225-H318-H336
DOTInformation : Class 3 - PG 2 - n-Propanol
EINECSNumber : 200-746-9
RTECSNumber : UH8225000