missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, puriss. p.a., ACS Reagent, ≥99.5% (GC), Solstice

Catalog No. 60046266
Change view
Click to view available options
Quantity:
1 L
2.5 L
6 x 1 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046266 1 L Poly Bottle
60046267 2.5 L Poly Bottle
60046269 6 x 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046266 Supplier Solstice Supplier No. 335381L

Puriss. p.a., Reag. Ph. Eur.,≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127°C
Color Colorless
Density 0.804g/mL at 25°C
Quantity 1 L
Explosion Limit 0.137
Flash Point 22°C (71.6°F)
Ignition Point 700°F
Odor Alcohol-like
Packaging Poly Bottle
Refractive Index n20/D 1.384
Vapor Pressure 14.9mmHg (20°C), 10mmHg (147°C)
Linear Formula CH3CH2CH2OH
UN Number UN1274
Beilstein 1098242
Boiling Point 97°C
Solubility Information Soluble in water and most organic solvents
Formula Weight 60.1g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material 1-Propanol
Show More Show Less
Hazard Statement P210-P261-P280-P305 + P351 + P338
DOTInformation Class 3 - PG 2 - n-Propanol
RTECSNumber 200-746-9