missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, HPLC Grade, 99% min

Catalog No. p-3256804
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA22932K2 1 L
AA22932M1 4 x 1 L
AA22932K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA22932K2 Supplier Thermo Scientific Chemicals Supplier No. 022932K2
Only null left

CAS: 71-23-8 | C3H8O | 60.10 g/mol

1-Propanol is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127°C
Density 0.804 g/cm3 at 20°C
Boiling Point 97°C
Flash Point 15°C (59°F)
Odor Alcohol-like
Assay Percent Range 99% min.
Refractive Index 1.385
Quantity 1 L
UN Number 1274
Beilstein 1098242
Solubility Information Miscible with water,alcohols,ketones,aldehydes,ethers,glycols,aromatic and aliphatic hydrocarbons.
Vapor Pressure 13.3 hPa (10mm Hg) at 20°C
Formula Weight 60.1
Grade HPLC
Physical Form Liquid
Chemical Name or Material 1-Propanol
Show More Show Less
Hazard Category H225-H318-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c
DOTInformation DOT Class: 3; Packing Group: II
EINECSNumber 200-746-9
RTECSNumber UH8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only