Learn More
1-Propanol (Certified), Fisher Chemical™
$247.75 - $584.08
Chemical Identifiers
| CAS | 71-23-8 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.10 |
| MDL Number | MFCD00002941 |
| InChI Key | BDERNNFJNOPAEC-UHFFFAOYSA-N |
| Synonym | 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol |
| PubChem CID | 1031 |
| ChEBI | CHEBI:28831 |
| IUPAC Name | propan-1-ol |
| SMILES | CCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
A414-500
|
Fisher Chemical
BPA414500 |
500 mL | HDPE Bottle |
Each for $247.75
Case of 6 Each for $1,486.50
|
|
||||
|
A414-1
|
Fisher Chemical
BPA4141 |
1 L | Glass Bottle |
Each for $326.92
Case of 6 Each for $1,961.52
|
|
||||
|
A414-4
|
Fisher Chemical
BPA4144 |
4 L | Glass Bottle |
Each for $584.08
Case of 4 Each for $2,336.32
|
|
||||
Chemical Identifiers
| 71-23-8 | |
| 60.10 | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| 1031 | |
| propan-1-ol |
| C3H8O | |
| MFCD00002941 | |
| 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
| CHEBI:28831 | |
| CCCO |
Specifications
| 7 | |
| 71-23-8 | |
| 0.02% max. | |
| 97°C | |
| Pass Test | |
| Liquid | |
| 0.801 to 0.803 (at 25°C) | |
| CH3CH2CH2OH | |
| 25mbar at 20°C | |
| 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
| CCCO | |
| 60.10 | |
| CHEBI:28831 | |
| Certified |
| -127°C | |
| Colorless | |
| 0.800g/cm³ | |
| 500 mL | |
| HDPE Bottle | |
| 0.01% max. | |
| C3H8O | |
| MFCD00002941 | |
| 2.2 mPa/s at 20°C | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| propan-1-ol | |
| 1031 | |
| 60.09 | |
| 1-Propanol |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause skin and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid