missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. aa41842k2
Change view
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41842K2 1 L
AA41842AK 250 mL
AA41842M1 4 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41842K2 Supplier Thermo Scientific Chemicals Supplier No. 041842K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71-23-8 | C3H8O | 60.10 g/mol

Used in making cosmetics, skin and hair preparations, pharmaceuticals, perfumes, lacquer formulations, dye solutions, antifreezes, rubbing alcohols, soaps, window cleaners, acetone and other chemicals and products. Also referred as ropyl alcohol;n-Propan-1-ol;n-Propanol;n-Propyl alcohol, Ethylcarbinol, Optal, Osmosol extra.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point °C to 127°C
Color Colorless
Appearance Clear liquid
Density 0.804 g/cm3 at 20°C
Quantity 1 L
Flash Point 15°C (59°F)
Ignition Point 824°C (1515°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.385
Assay Percent Range 99.9%
Vapor Pressure 13.3 hPa (10mm Hg) at 20°C
Beilstein 1098242
Boiling Point 97°C
Solubility Information Completely soluble in water. Miscible with propylene glycol.
Formula Weight 60.1
Chemical Name or Material 1-Propanol
Show More Show Less
Hazard Category H225-H318-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 200-746-9
RTECSNumber UH8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only