missing translation for 'onlineSavingsMsg'
Learn More

1-Propanethiol 98.0+%, TCI America™

Catalog No. p-7134632
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P048825ML 25 mL
P0488500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P048825ML Supplier TCI America Supplier No. P048825ML
Only null left

Chemical Identifiers

CAS 107-03-9
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.157
MDL Number MFCD00004900
InChI Key SUVIGLJNEAMWEG-UHFFFAOYSA-N
Synonym 1-propanethiol, propanethiol, n-propylmercaptan, propyl mercaptan, n-propyl mercaptan, n-propylthiol, 1-propylmercaptan, propylthiol, 1-mercaptopropane, 1-propyl mercaptan
PubChem CID 7848
ChEBI CHEBI:8473
IUPAC Name propane-1-thiol
SMILES CCCS

Specifications

Melting Point -113°C
Color Colorless
Boiling Point 67°C
Quantity 25 mL
UN Number 2402
Formula Weight 76.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Propanethiol
EINECSNumber (2)-0464
RTECSNumber TZ7300000
TSCA Yes