missing translation for 'onlineSavingsMsg'
Learn More

1-Propanethiol, 98%

CAS: 107-03-9 | C3H8S | 76.157 g/mol

$54.56 - $133.81

Chemical Identifiers

CAS 107-03-9
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.157
MDL Number MFCD00004900
InChI Key SUVIGLJNEAMWEG-UHFFFAOYSA-N
Synonym 1-propanethiol, propanethiol, n-propylmercaptan, propyl mercaptan, n-propyl mercaptan, n-propylthiol, 1-propylmercaptan, propylthiol, 1-mercaptopropane, 1-propyl mercaptan
PubChem CID 7848
ChEBI CHEBI:8473
IUPAC Name propane-1-thiol
SMILES CCCS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AA43420AE
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
043420AE
100 mL
Each for $54.56
Only null left
 
AA43420AP
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
043420AP
500 mL
Each for $133.81
Only null left
 
Description

Description

1-Propanethiol, is used widely in research areas. This compound has a wide range of applications including cell biology, materials science, nanoscience, self-assembled monolayers (SAMs), nanoelectronics, and contact printing. It is also used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Chemical Identifiers

107-03-9
76.157
SUVIGLJNEAMWEG-UHFFFAOYSA-N
7848
propane-1-thiol
C3H8S
MFCD00004900
1-propanethiol, propanethiol, n-propylmercaptan, propyl mercaptan, n-propyl mercaptan, n-propylthiol, 1-propylmercaptan, propylthiol, 1-mercaptopropane, 1-propyl mercaptan
CHEBI:8473
CCCS

Specifications

107-03-9
Colorless
67°C to 68°C
Stench
1.438
MFCD00004900
UN2402
1-propanethiol, propanethiol, n-propylmercaptan, propyl mercaptan, n-propyl mercaptan, n-propylthiol, 1-propylmercaptan, propylthiol, 1-mercaptopropane, 1-propyl mercaptan
SUVIGLJNEAMWEG-UHFFFAOYSA-N
propane-1-thiol
7848
76.16
Liquid
-113°C
0.8410 g/mL
−20°C (4°F)
C3H8S
CH3CH2CH2SH
100 mL
Air Sensitive
It is soluble in water (1.9g/L) at 25°C. It is also soluble in alcohol,acetone,benzene,ether,hydrocarbons.
CCCS
76.157
CHEBI:8473
98%
1-Propanethiol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H318-H302-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H225-H302-H315-H318-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPANETHIOLS

EINECSNumber : 203-455-5

RTECSNumber : TZ7300000

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only