missing translation for 'onlineSavingsMsg'
Learn More

1-Propanesulfonyl Chloride 98.0+%, TCI America™

Catalog No. P069325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P069325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P069325G Supplier TCI America Supplier No. P069325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10147-36-1
Molecular Formula C3H7ClO2S
Molecular Weight (g/mol) 142.597
MDL Number MFCD00007462
InChI Key KPBSJEBFALFJTO-UHFFFAOYSA-N
Synonym 1-propanesulfonyl chloride, propanesulfonyl chloride, propanesulphonyl chloride, propylsulfonyl chloride, 1-propanesulfonylchloride, n-propanesulfonyl chloride, 1-propane sulfonyl chloride, chloropropylsulfone, n-prso2cl, propanesulfonylchloride
PubChem CID 66279
IUPAC Name propane-1-sulfonyl chloride
SMILES CCCS(=O)(=O)Cl

Specifications

Color Yellow
Boiling Point 71°C
Quantity 25 g
UN Number 3265
Formula Weight 142.60
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Propanesulfonyl Chloride