missing translation for 'onlineSavingsMsg'
Learn More

1-Propanesulfonyl chloride, 97%

Catalog No. AAH60747AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60747AE 100 mL
AAH60747AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60747AE Supplier Thermo Scientific Chemicals Supplier No. H60747AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10147-36-1 | C3H7ClO2S | 142.597 g/mol

1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.

Solubility
Decomposes in hot water.

Notes
Moisture sensitive. Store in dry temperature.

Chemical Identifiers

CAS 10147-36-1
Molecular Formula C3H7ClO2S
Molecular Weight (g/mol) 142.597
MDL Number MFCD00007462
InChI Key KPBSJEBFALFJTO-UHFFFAOYSA-N
Synonym 1-propanesulfonyl chloride, propanesulfonyl chloride, propanesulphonyl chloride, propylsulfonyl chloride, 1-propanesulfonylchloride, n-propanesulfonyl chloride, 1-propane sulfonyl chloride, chloropropylsulfone, n-prso2cl, propanesulfonylchloride
PubChem CID 66279
IUPAC Name propane-1-sulfonyl chloride
SMILES CCCS(=O)(=O)Cl

Specifications

Density 1.267
Boiling Point 78°C to 79°C
Flash Point 97°C (207°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.453
Linear Formula CH3CH2CH2SO2Cl
Quantity 100 mL
UN Number UN3265
Beilstein 1747500
Sensitivity Moisture sensitive
Solubility Information Decomposes in hot water.
Formula Weight 142.6
Percent Purity 97%
Chemical Name or Material 1-Propanesulfonyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 233-414-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only