missing translation for 'onlineSavingsMsg'
Learn More

1-Piperonylpiperazine 97.0+%, TCI America™

Catalog No. P104125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P104125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P104125G Supplier TCI America Supplier No. P104125G
Only null left

Chemical Identifiers

CAS 32231-06-4
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD00005834
InChI Key NBOOZXVYXHATOW-UHFFFAOYSA-N
Synonym 1-piperonylpiperazine, 1-3,4-methylenedioxybenzyl piperazine, 1-benzo d 1,3 dioxol-5-ylmethyl piperazine, 1-1,3-benzodioxol-5-ylmethyl piperazine, 1-1,3-benzodioxol-5-yl-methyl piperazine, 1-benzo 1,3 dioxol-5-ylmethyl-piperazine, piperazine, 1-1,3-benzodioxol-5-ylmethyl, 1-3,4-methylendioxybenzyl piperazine, piperazine, 1-3,4-methylenedioxybenzyl, 5-piperazinylmethyl-2h-benzo d 1,3-dioxolene
PubChem CID 94426
IUPAC Name 1-(1,3-benzodioxol-5-ylmethyl)piperazine
SMILES C1CN(CCN1)CC2=CC3=C(C=C2)OCO3

Specifications

Melting Point 43°C
Boiling Point 149°C
Quantity 25 g
Formula Weight 220.27
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Piperonylpiperazine
RTECSNumber TM1550000
TSCA No