missing translation for 'onlineSavingsMsg'
Learn More

1-Piperidineethanol 98.0+%, TCI America™

Catalog No. p-7134567
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P034825G 25 g
P0348500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P034825G Supplier TCI America Supplier No. P034825G
Only null left

Chemical Identifiers

CAS 3040-44-6
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00006512
InChI Key KZTWONRVIPPDKH-UHFFFAOYSA-N
Synonym 2-piperidinoethanol, 1-piperidineethanol, 2-piperidin-1-yl ethanol, 1-2-hydroxyethyl piperidine, n-2-hydroxyethyl piperidine, n-piperidineethanol, 2-1-piperidinyl ethanol, n-hydroxyethyl piperidine, beta-piperidylethanol, beta-piperidinoethanol
PubChem CID 18232
ChEBI CHEBI:61238
IUPAC Name 2-piperidin-1-ylethanol
SMILES C1CCN(CC1)CCO

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 g
Formula Weight 129.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Piperidineethanol
RTECSNumber TM8052000
TSCA Yes