missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylsulfonyl)-1H-indole-2-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 40899-93-2
Molecular Formula C15H11NO4S
Molecular Weight (g/mol) 301.316
MDL Number MFCD03086089
InChI Key QIWDUGKJAHJRAE-UHFFFAOYSA-N
Synonym 1-phenylsulfonyl-1h-indole-2-carboxylic acid, 1-benzenesulfonyl indole-2-carboxylic acid, 1-benzenesulfonyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylicacid, 1-phenylsulfonyl
PubChem CID 2776219
IUPAC Name 1-(benzenesulfonyl)indole-2-carboxylic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)N2C3=CC=CC=C3C=C2C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC03301DA
SDS View Documents
Thermo Scientific Chemicals
CC03301DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC03301DE
SDS View Documents
Thermo Scientific Chemicals
CC03301DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

40899-93-2
301.316
QIWDUGKJAHJRAE-UHFFFAOYSA-N
2776219
C1=CC=C(C=C1)S(=O)(=O)N2C3=CC=CC=C3C=C2C(=O)O
C15H11NO4S
MFCD03086089
1-phenylsulfonyl-1h-indole-2-carboxylic acid, 1-benzenesulfonyl indole-2-carboxylic acid, 1-benzenesulfonyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylicacid, 1-phenylsulfonyl
1-(benzenesulfonyl)indole-2-carboxylic acid

Specifications

40899-93-2
100.0
Amber glass bottle
MFCD03086089
1-phenylsulfonyl-1h-indole-2-carboxylic acid, 1-benzenesulfonyl indole-2-carboxylic acid, 1-benzenesulfonyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylicacid, 1-phenylsulfonyl
C1=CC=C(C=C1)S(=O)(=O)N2C3=CC=CC=C3C=C2C(=O)O
301.316
301.32
1-(Phenylsulfonyl)-1H-indole-2-carboxylic acid
96.5
97%
C15H11NO4S
1g
QIWDUGKJAHJRAE-UHFFFAOYSA-N
1-(benzenesulfonyl)indole-2-carboxylic acid
2776219
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'