missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylsemicarbazide 98.0+%, TCI America™

Catalog No. p022725g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P022725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P022725G Supplier TCI America Supplier No. P022725G
Only null left

Chemical Identifiers

CAS 103-03-7
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007942
InChI Key AVKHCKXGKPAGEI-UHFFFAOYSA-N
Synonym phenicarbazide, 1-phenylsemicarbazide, 2-phenylhydrazinecarboxamide, phenicarbazid, phenylsemicarbazide, carbaphen, kryogenin, phenygenine, febrimin, 1-carbamyl-2-phenylhydrazine
PubChem CID 61002
ChEBI CHEBI:82521
IUPAC Name (phenylamino)urea
SMILES NC(=O)NNC1=CC=CC=C1

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenylsemicarbazide
RTECSNumber FD0575000
TSCA Yes