missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpyrrolidine, 98+%

Catalog No. AAL0523906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0523906 5 g
AAL0523914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0523906 Supplier Thermo Scientific Chemicals Supplier No. L0523906

CAS: 4096-21-3 | C10H13N | 147.22 g/mol

1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents. Store cold 2-8°C.

Chemical Identifiers

CAS 4096-21-3
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00015897
InChI Key VDQQJMHXZCMNMU-UHFFFAOYSA-N
Synonym 1-phenyl-pyrrolidine, n-phenylpyrrolidine, phenylpyrrolidine, pyrrolidine, 1-phenyl, 1-phenyl-pyrrolidin-3-ol, 1-phenylpyrolidine, n-phenyl pyrrolidine, pubchem9192, acmc-1cttc
PubChem CID 77726
IUPAC Name 1-phenylpyrrolidine
SMILES C1CCN(C1)C1=CC=CC=C1

Specifications

Boiling Point 133°C to 134°C (19 mmHg)
Refractive Index 1.582
Quantity 5 g
Beilstein 117030
Solubility Information Not miscible in water.
Formula Weight 147.22
Percent Purity ≥98%
Chemical Name or Material 1-Phenylpyrrolidine
Hazard Category H302+H312
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 223-849-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only