missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpyrrole, 99%

Catalog No. AAL0797306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0797306 5 g
AAL0797314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0797306 Supplier Thermo Scientific Chemicals Supplier No. L0797306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 635-90-5 | C10H9N | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 635-90-5
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00005343
InChI Key GEZGAZKEOUKLBR-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrrole, n-phenylpyrrole, pyrrole, 1-phenyl, 1h-pyrrole, 1-phenyl, unii-3od6n6545h, chembl84459, 1-phenylpyrrol, acmc-209ngl, 1-phenylpyrrole, 1-phenyl-1h-pyrrole #
PubChem CID 12480
IUPAC Name 1-phenylpyrrole
SMILES C1=CN(C=C1)C1=CC=CC=C1

Specifications

Melting Point 57°C to 60°C
Boiling Point 131°C (30 mmHg)
Quantity 5 g
Beilstein 109798
Formula Weight 143.19
Percent Purity 99%
Chemical Name or Material 1-Phenylpyrrole
Hazard Category H315-H320-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-242-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only