missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpiperazine 98.0+%, TCI America™

Catalog No. P098025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P098025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P098025G Supplier TCI America Supplier No. P098025G

Specifications

CAS 92-54-6
Color Yellow
Boiling Point 272°C
Molecular Formula C10H14N2
Quantity 25 g
MDL Number MFCD00005957
UN Number 2922
Synonym phenylpiperazine, n-phenylpiperazine, piperazine, 1-phenyl, 1-phenyl-piperazine, n-phenyldiethylenediamine, 1-fenylpiperazin czech, unii-j9225cbi7d, n-phenyl piperazine, 1-phenyl piperazine, ccris 4334
InChI Key YZTJYBJCZXZGCT-UHFFFAOYSA-N
SMILES C1CN(CCN1)C2=CC=CC=C2
IUPAC Name 1-phenylpiperazine
Molecular Weight (g/mol) 162.236
PubChem CID 7096
Formula Weight 162.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenylpiperazine
Show More Show Less
EINECSNumber (5)-0958
RTECSNumber TM2625000
TSCA Yes