missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpiperazine hydrochloride, 99%

Catalog No. AAA1699514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1699514 25 g
AAA1699522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1699514 Supplier Thermo Scientific Chemicals Supplier No. A1699514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2210-93-7 | C10H15ClN2 | 198.694 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2210-93-7
Molecular Formula C10H15ClN2
Molecular Weight (g/mol) 198.694
MDL Number MFCD00012758
InChI Key NQNZNONJZASOKL-UHFFFAOYSA-N
Synonym 1-phenylpiperazine hydrochloride, 1-phenylpiperazinium chloride, piperazine, 1-phenyl-, hydrochloride, phenylpiperazine hydrochloride, 1-phenyl piperazine hydrochloride, piperazine, 1-phenyl-, monohydrochloride, phenylpiperazine, chloride, 1-phenylpiperazine hcl, c10h14n2.hcl, 1-phenylpiperazinehydrochloride
PubChem CID 75164
IUPAC Name 1-phenylpiperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=CC=C2.Cl

Specifications

Melting Point 247°C to 250°C
Odor Odorless
Quantity 25 g
UN Number UN2811
Formula Weight 198.7
Percent Purity 99%
Chemical Name or Material 1-Phenylpiperazine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 218-646-9
RTECSNumber TM2640000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only