Learn More
1-Phenylpiperazine, 98+%
CAS: 92-54-6 | C10H14N2 | 162.236 g/mol
Supplier: Thermo Scientific Chemicals A1261022
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Phenylpiperazine | |
| 17°C to 19°C | |
| 285°C to 286°C | |
| C10H14N2 | |
| MFCD00005957 | |
| UN2927 | |
| phenylpiperazine, n-phenylpiperazine, piperazine, 1-phenyl, 1-phenyl-piperazine, n-phenyldiethylenediamine, 1-fenylpiperazin czech, unii-j9225cbi7d, n-phenyl piperazine, 1-phenyl piperazine, ccris 4334 | |
| C1CN(CCN1)C2=CC=CC=C2 | |
| 162.236 | |
| 162.24 |
| 92-54-6 | |
| 1.064 | |
| 141°C (286°F) | |
| 1.588 | |
| 100 g | |
| 132157 | |
| YZTJYBJCZXZGCT-UHFFFAOYSA-N | |
| 1-phenylpiperazine | |
| 7096 | |
| ≥98% |
Chemical Identifiers
| 92-54-6 | |
| 162.236 | |
| YZTJYBJCZXZGCT-UHFFFAOYSA-N | |
| 7096 | |
| C1CN(CCN1)C2=CC=CC=C2 |
| C10H14N2 | |
| MFCD00005957 | |
| phenylpiperazine, n-phenylpiperazine, piperazine, 1-phenyl, 1-phenyl-piperazine, n-phenyldiethylenediamine, 1-fenylpiperazin czech, unii-j9225cbi7d, n-phenyl piperazine, 1-phenyl piperazine, ccris 4334 | |
| 1-phenylpiperazine |
Safety and Handling
GHS H Statement
H301-H310-H314-H318
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H310-H314-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber : 202-165-6
RTECSNumber : TM2625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only