missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpiperazine, 98+%

Catalog No. AAA1261014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1261014 25 g
AAA1261022 100 g
AAA1261036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1261014 Supplier Thermo Scientific Chemicals Supplier No. A1261014

CAS: 92-54-6 | C10H14N2 | 162.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-54-6
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
MDL Number MFCD00005957
InChI Key YZTJYBJCZXZGCT-UHFFFAOYSA-N
Synonym phenylpiperazine, n-phenylpiperazine, piperazine, 1-phenyl, 1-phenyl-piperazine, n-phenyldiethylenediamine, 1-fenylpiperazin czech, unii-j9225cbi7d, n-phenyl piperazine, 1-phenyl piperazine, ccris 4334
PubChem CID 7096
IUPAC Name 1-phenylpiperazine
SMILES C1CN(CCN1)C2=CC=CC=C2

Specifications

Melting Point 17°C to 19°C
Density 1.064
Boiling Point 285°C to 286°C
Flash Point 141°C (286°F)
Refractive Index 1.588
Quantity 25 g
UN Number UN2927
Beilstein 132157
Formula Weight 162.24
Percent Purity ≥98%
Chemical Name or Material 1-Phenylpiperazine
Show More Show Less
Hazard Category H301-H310-H314-H335
Hazard Statement GHS H Statement
H301-H310-H314-H318
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 202-165-6
RTECSNumber TM2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only