missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylnaphthalene 97.0+%, TCI America™

Catalog No. P06325G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P06325G Supplier TCI America Supplier No. P06325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 605-02-7
Molecular Formula C16H12
Molecular Weight (g/mol) 204.27
MDL Number MFCD00003983
InChI Key IYDMICQAKLQHLA-UHFFFAOYSA-N
Synonym naphthalene, 1-phenyl, 1-phenyl-naphthalene, unii-10nxc4g45q, phenylnaphthalene, phenyl-napthalene, p-4-naphthyl benzene, acmc-1bfcg, 1-phenylnaphthalene, naphthalene, 1-phenyl-8ci 9ci, 10 microg/ml in acetonitrile
PubChem CID 11795
IUPAC Name 1-phenylnaphthalene
SMILES C1=CC=C(C=C1)C1=C2C=CC=CC2=CC=C1

Specifications

Color Yellow
Boiling Point 320°C
Quantity 5 g
Formula Weight 204.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenylnaphthalene