missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylnaphthalene, 97%

Catalog No. AAB2195306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2195306 5 g
AAB2195314 25 g
AAB2195322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2195306 Supplier Thermo Scientific Chemicals Supplier No. B2195306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 605-02-7 | C16H12 | 204.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 605-02-7
Molecular Formula C16H12
Molecular Weight (g/mol) 204.27
MDL Number MFCD00003983
InChI Key IYDMICQAKLQHLA-UHFFFAOYSA-N
Synonym naphthalene, 1-phenyl, 1-phenyl-naphthalene, unii-10nxc4g45q, phenylnaphthalene, phenyl-napthalene, p-4-naphthyl benzene, acmc-1bfcg, 1-phenylnaphthalene, naphthalene, 1-phenyl-8ci 9ci, 10 microg/ml in acetonitrile
PubChem CID 11795
IUPAC Name 1-phenylnaphthalene
SMILES C1=CC=C(C=C1)C1=C2C=CC=CC2=CC=C1

Specifications

Density 1.088
Boiling Point 320°C
Flash Point >110°C (230°F)
Refractive Index 1.664
Quantity 5 g
Beilstein 1906885
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 1-Phenylnaphthalene
EINECSNumber 210-081-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only