missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylisatin, 98%

Catalog No. AAL1138806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1138806 5 g
AAL1138803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1138806 Supplier Thermo Scientific Chemicals Supplier No. L1138806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 723-89-7 | C14H9NO2 | 223.231 g/mol

1-Phenylisatin is a reactant for preparation of indolin-2,3-dione Schiff bases as potential antimycobacterial agents, antipyretic agents, antiproliferative agents, phenylacetamides as anticonvulsant agents and selective Galanin GAL3 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenylisatin is a reactant for preparation of indolin-2,3-dione Schiff bases as potential antimycobacterial agents, antipyretic agents, antiproliferative agents, phenylacetamides as anticonvulsant agents and selective Galanin GAL3 receptor antagonists.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 723-89-7
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.231
MDL Number MFCD00082681
InChI Key UWCPWBIMRYXUOU-UHFFFAOYSA-N
Synonym 1-phenylisatin, 1-phenylindoline-2,3-dione, 1-phenyl-1h-indole-2,3-dione, 1h-indole-2,3-dione, 1-phenyl, 1-phenyl-indole-2,3-dione, indole-2,3-dione, 1-phenyl, 1h-indole-2,3-dione, 1-phenyl-9ci, 1-phenyl-2,3-dihydro-1h-indole-2,3-dione, phenylisatin, n-phenylisatin
PubChem CID 12884
IUPAC Name 1-phenylindole-2,3-dione
SMILES C1=CC=C(C=C1)N2C3=CC=CC=C3C(=O)C2=O

Specifications

Melting Point 139°C to 142°C
Quantity 5 g
Beilstein 164531
Solubility Information Insoluble in water.
Formula Weight 223.23
Percent Purity 98%
Chemical Name or Material 1-Phenylisatin
EINECSNumber 000-000-0
RTECSNumber NL7984000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only