missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylimidazole 98.0+%, TCI America™

Catalog No. P20305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20305G 5 g
P20301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20305G Supplier TCI America Supplier No. P20305G
Only null left

Chemical Identifiers

CAS 7164-98-9
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00041204
InChI Key SEULWJSKCVACTH-UHFFFAOYSA-N
Synonym 1-phenyl-1h-imidazole, phenylimidazole, n-phenylimidazole, 1h-imidazole, 1-phenyl, 1h-imidazole, phenyl, n-phenyl imidazole, pubchem12611, acmc-20aos8, 1-phenylimidazole, 1-phenyl-1h-imidazole #
PubChem CID 81595
IUPAC Name 1-phenyl-1H-imidazole
SMILES C1=CN(C=N1)C1=CC=CC=C1

Specifications

Melting Point 13°C
Color Yellow
Boiling Point 142°C
Quantity 5 g
Formula Weight 144.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenylimidazole