missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenylimidazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 13°C |
| Color | Yellow |
| Boiling Point | 142°C |
| Quantity | 1 g |
| Formula Weight | 144.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Phenylimidazole |
Safety and Handling
| TSCA | No |