missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Phenylethylamine, 98+%

Catalog No. AAA15711AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15711AP 500 mL
AAA15711AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA15711AP Supplier Thermo Scientific Chemicals Supplier No. A15711AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 618-36-0 | C8H11N | 121.183 g/mol

Used for synthesis and as an emulsifying agent; used as a resolving agent and chiral intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for synthesis and as an emulsifying agent; used as a resolving agent and chiral intermediate.

Solubility
Soluble in water at 20°C 4.2 g/L.

Notes
Air sensitive. Store away from air, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.

Chemical Identifiers

CAS 618-36-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008069
InChI Key RQEUFEKYXDPUSK-UHFFFAOYSA-N
Synonym 1-phenylethylamine, alpha-methylbenzylamine, alpha-phenylethylamine, dl-alpha-methylbenzylamine, 1-phenethylamine, alpha-aminoethylbenzene, 1-amino-1-phenylethane, 1-phenylethan-1-amine, a-methylbenzylamine, alpha-methylbenzenemethanamine
PubChem CID 7408
ChEBI CHEBI:670
IUPAC Name 1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Density 0.945
Boiling Point 69°C to 70°C (10 mmHg)
Flash Point 79°C (174°F)
Odor Odorless
Refractive Index 1.527
Quantity 500 mL
UN Number UN2735
Beilstein 636127
Sensitivity Air Sensitive
Merck Index 14,6026
Solubility Information Soluble in water at 20°C 4.2g/L.
Formula Weight 121.18
Percent Purity ≥98%
Chemical Name or Material (+/-)-1-Phenylethylamine
Show More Show Less
Hazard Category H227-H302+H312-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 210-545-8
RTECSNumber DP5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only