missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Phenylethyl Alcohol 98.0+%, TCI America™

$67.11 - $184.54

Chemical Identifiers

CAS 98-85-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004508
InChI Key WAPNOHKVXSQRPX-UHFFFAOYSA-N
Synonym methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene
PubChem CID 7409
ChEBI CHEBI:669
IUPAC Name 1-phenylethanol
SMILES CC(C1=CC=CC=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M016325ML
View Documents
TCI America
M016325ML
25 mL
Each for $67.11
Only null left
 
M0163500ML
View Documents
TCI America
M0163500ML
500 mL
Each for $184.54
Only null left
 
Description

Description

Specifications

Chemical Identifiers

98-85-1
122.167
WAPNOHKVXSQRPX-UHFFFAOYSA-N
7409
1-phenylethanol
C8H10O
MFCD00004508
methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene
CHEBI:669
CC(C1=CC=CC=C1)O

Specifications

98-85-1
205°C
MFCD00004508
2937
WAPNOHKVXSQRPX-UHFFFAOYSA-N
1-phenylethanol
7409
122.17
Liquid
Colorless
C8H10O
25 mL
methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene
CC(C1=CC=CC=C1)O
122.167
CHEBI:669
≥98.0% (GC)
(+/-)-1-Phenylethyl Alcohol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1012&(3)-1049

RTECSNumber : DO9275000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'