Learn More
(+/-)-1-Phenylethanol, 97%
CAS: 98-85-1 | C8H10O | 122.167 g/mol
$85.45 - $85.45
Chemical Identifiers
| CAS | 98-85-1 |
|---|---|
| Molecular Formula | C8H10O |
| Molecular Weight (g/mol) | 122.167 |
| MDL Number | MFCD00004508 |
| InChI Key | WAPNOHKVXSQRPX-UHFFFAOYSA-N |
| Synonym | methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene |
| PubChem CID | 7409 |
| ChEBI | CHEBI:669 |
| IUPAC Name | 1-phenylethanol |
| SMILES | CC(C1=CC=CC=C1)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1383736
|
Thermo Scientific Chemicals
A1383736 |
500 g |
Each for $85.45
|
|
|||||
Description
The Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
Chemical Identifiers
| 98-85-1 | |
| 122.167 | |
| WAPNOHKVXSQRPX-UHFFFAOYSA-N | |
| 7409 | |
| 1-phenylethanol |
| C8H10O | |
| MFCD00004508 | |
| methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene | |
| CHEBI:669 | |
| CC(C1=CC=CC=C1)O |
Specifications
| 98-85-1 | |
| 1.012 | |
| C8H10O | |
| 500 g | |
| methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene | |
| WAPNOHKVXSQRPX-UHFFFAOYSA-N | |
| 1-phenylethanol | |
| 7409 | |
| 122.17 | |
| (+/-)-1-Phenylethanol |
| 19°C to 20°C | |
| 203°C to 205°C | |
| MFCD00004508 | |
| UN2937 | |
| Insoluble in water. | |
| CC(C1=CC=CC=C1)O | |
| 122.167 | |
| CHEBI:669 | |
| 97% |
Safety and Handling
GHS H Statement
H318-H302-H335-H227-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
P210-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
H227-H302-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALPHA-METHYLBENZYL ALCOHOL, LIQUID
EINECSNumber : 202-707-1
RTECSNumber : DO9275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only