missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylethane-1,2-diol 98.0+%, TCI America™

Catalog No. p-7134729
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P068625G 25 g
P0686250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P068625G Supplier TCI America Supplier No. P068625G
Only null left

Chemical Identifiers

CAS 93-56-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00003546
InChI Key PWMWNFMRSKOCEY-UHFFFAOYSA-N
Synonym 1-phenyl-1,2-ethanediol, styrene glycol, phenylethylene glycol, phenylethanediol, phenyl glycol, 1,2-ethanediol, 1-phenyl, phenyl-1,2-ethanediol, styrolyl alcohol, fenylglycol, 1-phenylethylene glycol
PubChem CID 7149
IUPAC Name 1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O

Specifications

Melting Point 67°C
Color White-Yellow
Boiling Point 82°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenylethane-1,2-diol
EINECSNumber (3)-2882
RTECSNumber KI2500000
TSCA Yes